Learn More
Waters Corp Tubing Assembly, MP35N, 0.062 in. x 0.004 in. x 27.0 in., High Pressure
Catalog No. 502390469 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed for high-pressure applications and is compatible with H-Class Bio Systems. It features MP35N material and specific dimensions for reliable performance.
  • Constructed from MP35N material.
  • Features an outer diameter of 0.062 in.
  • Internal diameter of 0.004 in.
  • Length of 27 in.
  • Provided with high pressure fittings.
  • Designed for UPLC systems.
Catalog No. 50-239-0469 Supplier Waters Corp Supplier No. 430002974
This tubing assembly is designed for high-pressure applications and is compatible with H-Class Bio Systems. It features MP35N material and specific dimensions for reliable performance.
  • Constructed from MP35N material.
  • Features an outer diameter of 0.062 in.
  • Internal diameter of 0.004 in.
  • Length of 27 in.
  • Provided with high pressure fittings.
  • Designed for UPLC systems.