Learn More
Waters Corp Quality Parts Tubing Assembly, MP35N, 0.004 in. ID, 22.5 in.
Catalog No. 502391865 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement tubing assembly is constructed from MP35N material. It features precise dimensions of 0.062 in. x 0.004 in. x 22.5 in. and includes low pressure (LP) fittings, making it suitable for systems with a CH-30A.
  • Provided with low pressure fittings.
  • Found on systems with a CH-30A.
Catalog No. 50-239-1865 Supplier Waters Corp Supplier No. 700008914
This replacement tubing assembly is constructed from MP35N material. It features precise dimensions of 0.062 in. x 0.004 in. x 22.5 in. and includes low pressure (LP) fittings, making it suitable for systems with a CH-30A.
  • Provided with low pressure fittings.
  • Found on systems with a CH-30A.