Learn More
Waters Corp Tubing Assembly, MP35N, 0.004 in. ID, High Pressure
Catalog No. 502392073 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is a replacement component provided with high pressure fittings and is found on H-Class Bio CM-A Systems.
  • Provided with high pressure fittings
  • Made from MP35N material
  • Internal diameter of 0.004 in.
  • Length of 14.5 in.
  • Compatible with UPLC system type
Catalog No. 50-239-2073 Supplier Waters Corp Supplier No. 700005481
This tubing assembly is a replacement component provided with high pressure fittings and is found on H-Class Bio CM-A Systems.
  • Provided with high pressure fittings
  • Made from MP35N material
  • Internal diameter of 0.004 in.
  • Length of 14.5 in.
  • Compatible with UPLC system type