Learn More
Waters Corp Tubing Assembly, MP35N, 0.004 in. ID, 19.0 in. Length
Catalog No. 502392888 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is a replacement part provided with high pressure fittings. It features MP35N material with a 0.004 in. internal diameter and 19.0 in. length, making it suitable for use in H-Class Bio CM-A Systems.
  • Manufactured from MP35N material.
  • Features a 0.004 in. internal diameter.
  • Measures 19 in. in length.
  • Provided with high pressure fittings.
  • Designed for UPLC systems.
  • Compatible with H-Class Bio CM-A systems.
  • Supplied as 1 unit per package.
Catalog No. 50-239-2888 Supplier Waters Corp Supplier No. 700005484
This tubing assembly is a replacement part provided with high pressure fittings. It features MP35N material with a 0.004 in. internal diameter and 19.0 in. length, making it suitable for use in H-Class Bio CM-A Systems.
  • Manufactured from MP35N material.
  • Features a 0.004 in. internal diameter.
  • Measures 19 in. in length.
  • Provided with high pressure fittings.
  • Designed for UPLC systems.
  • Compatible with H-Class Bio CM-A systems.
  • Supplied as 1 unit per package.