Learn More
Waters Corp Tubing Assembly, MP35N, GPV to Primary Check Valve, UPLC System
Catalog No. 502391469 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is a replacement part for an ACQUITY Arc Bio Quaternary Solvent Manager. It features MP35N material and is designed to connect the GPV to the primary check valve.
  • Tubing type
  • UPLC system compatible
  • Made from MP35N material
  • Designed for replacement as required
  • For ACQUITY Arc Bio Quaternary Solvent Manager
Catalog No. 50-239-1469 Supplier Waters Corp Supplier No. 700011677
This tubing assembly is a replacement part for an ACQUITY Arc Bio Quaternary Solvent Manager. It features MP35N material and is designed to connect the GPV to the primary check valve.
  • Tubing type
  • UPLC system compatible
  • Made from MP35N material
  • Designed for replacement as required
  • For ACQUITY Arc Bio Quaternary Solvent Manager