Learn More
Waters Corp Tubing Assembly, MP35N, Pre-Heater to Column, Reusable
Catalog No. 502391749 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement tubing assembly is constructed from MP35N material and is designed to connect the pre-heater to the column. It is reusable and intended for use with a CH-30A column heater in a bio-compatible system.
  • Made from MP35N material
  • Connects pre-heater to column
  • Reusable
  • Designed for UPLC systems
  • Intended for use with a CH-30A column heater in bio-compatible systems
Catalog No. 50-239-1749 Supplier Waters Corp Supplier No. 700008918
This replacement tubing assembly is constructed from MP35N material and is designed to connect the pre-heater to the column. It is reusable and intended for use with a CH-30A column heater in a bio-compatible system.
  • Made from MP35N material
  • Connects pre-heater to column
  • Reusable
  • Designed for UPLC systems
  • Intended for use with a CH-30A column heater in bio-compatible systems