Learn More
Waters Corp Tubing Assembly, MP35N, Pre-Heater to Column, UPLC System
Catalog No. 502391573 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is a replacement part for use with a CH-30A column heater in a Bio-compatible system. It connects the pre-heater to the column.
  • Designed for UPLC systems.
  • Constructed from MP35N material.
  • Connects pre-heater to column.
Catalog No. 50-239-1573 Supplier Waters Corp Supplier No. 700008917
This tubing assembly is a replacement part for use with a CH-30A column heater in a Bio-compatible system. It connects the pre-heater to the column.
  • Designed for UPLC systems.
  • Constructed from MP35N material.
  • Connects pre-heater to column.