Learn More
Waters Corp Tubing Assembly, PEEK, 0.062 in. OD, 0.020 in. ID, 60 in. Length
Catalog No. 502392934 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement tubing assembly is constructed from PEEK material, featuring dimensions of 0.062 in. outer diameter, 0.020 in. internal diameter, and a length of 60 in. It functions as the waste line for ACQUITY Arc systems.
  • PEEK material construction.
  • Dimensions: 0.062 in. outer diameter, 0.020 in. internal diameter, and 60 in. length.
  • Functions as the waste line in ACQUITY Arc systems.
  • Product type is tubing.
  • Suitable for UPLC system types.
  • Packaged with 1 unit per package.
  • Replacement schedule is “As Required.”
Catalog No. 50-239-2934 Supplier Waters Corp Supplier No. 700010531
This replacement tubing assembly is constructed from PEEK material, featuring dimensions of 0.062 in. outer diameter, 0.020 in. internal diameter, and a length of 60 in. It functions as the waste line for ACQUITY Arc systems.
  • PEEK material construction.
  • Dimensions: 0.062 in. outer diameter, 0.020 in. internal diameter, and 60 in. length.
  • Functions as the waste line in ACQUITY Arc systems.
  • Product type is tubing.
  • Suitable for UPLC system types.
  • Packaged with 1 unit per package.
  • Replacement schedule is “As Required.”