Learn More
Waters Corp Tubing Assembly, PEEK, Solvent Manager Purge Line, 0.04 in. ID
Catalog No. 502392411 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is a replacement part made of PEEK, designed specifically as a purge line for the Solvent Manager on ACQUITY Arc and Arc HPLC Systems.
  • Used on ACQUITY Arc and Arc HPLC Systems
  • Constructed from PEEK material
  • Internal diameter of 0.04 in.
  • Length of 60 in.
  • Functions as a purge line for the solvent manager
  • Suitable for UPLC system type
Catalog No. 50-239-2411 Supplier Waters Corp Supplier No. 700010544
This tubing assembly is a replacement part made of PEEK, designed specifically as a purge line for the Solvent Manager on ACQUITY Arc and Arc HPLC Systems.
  • Used on ACQUITY Arc and Arc HPLC Systems
  • Constructed from PEEK material
  • Internal diameter of 0.04 in.
  • Length of 60 in.
  • Functions as a purge line for the solvent manager
  • Suitable for UPLC system type