Learn More
Waters Corp Waters Tubing Assembly, Pre-Heater to Column, Reusable, MP35N
Catalog No. 502224481 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed for use in BioACQUITY systems, connecting an APH to the analytical column within a CH-30A column heater. It is a reusable component made from MP35N material, ensuring reliability and compatibility for analytical applications.
  • Connects pre-heater to column
  • Reusable design
  • Constructed from MP35N material
  • Suitable for UPLC system types
  • Internal diameter of 0.004 in.
  • Replaced as required to maintain system performance.
Catalog No. 50-222-4481 Supplier Waters Corp Supplier No. 700011809
This tubing assembly is designed for use in BioACQUITY systems, connecting an APH to the analytical column within a CH-30A column heater. It is a reusable component made from MP35N material, ensuring reliability and compatibility for analytical applications.
  • Connects pre-heater to column
  • Reusable design
  • Constructed from MP35N material
  • Suitable for UPLC system types
  • Internal diameter of 0.004 in.
  • Replaced as required to maintain system performance.