Learn More
Waters Corp Tubing Assembly, Sample Loading, Inlet Manifold Valve Connection
Catalog No. 502391559 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed for sample loading, providing a connection to the inlet manifold valve for direct sample loading through the pump.
  • Connects to left side of inlet manifold valve
  • Used for direct sample loading through pump
Catalog No. 50-239-1559 Supplier Waters Corp Supplier No. 430002144
This tubing assembly is designed for sample loading, providing a connection to the inlet manifold valve for direct sample loading through the pump.
  • Connects to left side of inlet manifold valve
  • Used for direct sample loading through pump