Learn More
Waters Corp Tubing Assembly, Solenoid A to Waste, Stainless Steel, 0.04 in.
Catalog No. 502392285 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is a replacement part for the Solenoid A to Waste connection, specifically associated with 2414 Refractive Index Detectors in HPLC systems. It is constructed from durable stainless steel.
  • Designed as a replacement part for Solenoid A to Waste
  • Associated with 2414 Refractive Index Detectors
  • Suitable for HPLC systems
  • Constructed from stainless steel
  • Features an internal diameter of 0.04 in.
  • Measures 20 in. in length
  • Includes 1 unit per package
Catalog No. 50-239-2285 Supplier Waters Corp Supplier No. 700002377
This tubing assembly is a replacement part for the Solenoid A to Waste connection, specifically associated with 2414 Refractive Index Detectors in HPLC systems. It is constructed from durable stainless steel.
  • Designed as a replacement part for Solenoid A to Waste
  • Associated with 2414 Refractive Index Detectors
  • Suitable for HPLC systems
  • Constructed from stainless steel
  • Features an internal diameter of 0.04 in.
  • Measures 20 in. in length
  • Includes 1 unit per package