Learn More
Waters Corp Tubing Assembly, Solvent Filter Adapter, UPLC
Catalog No. 502391222 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement tubing assembly provides connectivity to Solvent Filter 700003615 (which is supplied separately) for ACQUITY Arc and Arc HPLC Sample Manager FTNs.
  • Connects to solvent filters.
  • Suitable for UPLC systems.
  • Functions as a tubing component.
  • Recommended for replacement as required.
Catalog No. 50-239-1222 Supplier Waters Corp Supplier No. 700010541
This replacement tubing assembly provides connectivity to Solvent Filter 700003615 (which is supplied separately) for ACQUITY Arc and Arc HPLC Sample Manager FTNs.
  • Connects to solvent filters.
  • Suitable for UPLC systems.
  • Functions as a tubing component.
  • Recommended for replacement as required.