Learn More
Waters Corp Tubing Assembly, SSV to i2V, 1/16 in. OD, 0.040 in. ID
Catalog No. 502391571 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly connects the Solvent Select Valve (SSV) to the Intelligent Intake Valve (I2V). The Intelligent Intake Valve is a component found in the ACQUITY Binary Solvent Manager and the ACQUITY I-Class BSM.
  • Connects the Solvent Select Valve (SSV) to the Intelligent Intake Valve (I2V)
  • Component of the ACQUITY Binary Solvent Manager and ACQUITY I-Class BSM
  • Outer diameter of 1/16 in.
  • Inner diameter of 0.040 in.
  • Supplied as a spare parts kit
  • Contains 1 unit per package
  • Replacement schedule as required
Catalog No. 50-239-1571 Supplier Waters Corp Supplier No. 430001443
This tubing assembly connects the Solvent Select Valve (SSV) to the Intelligent Intake Valve (I2V). The Intelligent Intake Valve is a component found in the ACQUITY Binary Solvent Manager and the ACQUITY I-Class BSM.
  • Connects the Solvent Select Valve (SSV) to the Intelligent Intake Valve (I2V)
  • Component of the ACQUITY Binary Solvent Manager and ACQUITY I-Class BSM
  • Outer diameter of 1/16 in.
  • Inner diameter of 0.040 in.
  • Supplied as a spare parts kit
  • Contains 1 unit per package
  • Replacement schedule as required