Learn More
Waters Corp Tubing Assembly, Stainless Steel, 0.005 in ID, Valve Inlet
Catalog No. 502392979 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is constructed from stainless steel with a 0.005 in internal diameter. It is designed for use on the 3 Column Select Valve in ACQUITY Arc Systems.
  • Constructed from stainless steel
  • Features a 0.005 in internal diameter
  • Designed for use on the 3 Column Select Valve in ACQUITY Arc Systems
  • Type: Tubing
  • System type: UPLC
Catalog No. 50-239-2979 Supplier Waters Corp Supplier No. 700010694
This tubing assembly is constructed from stainless steel with a 0.005 in internal diameter. It is designed for use on the 3 Column Select Valve in ACQUITY Arc Systems.
  • Constructed from stainless steel
  • Features a 0.005 in internal diameter
  • Designed for use on the 3 Column Select Valve in ACQUITY Arc Systems
  • Type: Tubing
  • System type: UPLC