Learn More
Waters Corp Tubing Assembly, Stainless Steel, 0.062 in. OD, 0.004 in. ID, 44.0 in. Length
Catalog No. 502392793 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is a stainless steel spare part, designed for high-pressure applications. It features specific internal and external diameters and is supplied with high pressure fittings, suitable for use in ACQUITY H-Class Systems.
  • Made from stainless steel.
  • Has an internal diameter of 0.004 in. (0.102 mm).
  • Measures 44.0 in. (1118 mm) in length.
  • Supplied with high pressure fittings.
  • Designed for UPLC systems.
  • Suitable for use in ACQUITY H-Class Systems.
Catalog No. 50-239-2793 Supplier Waters Corp Supplier No. 430002992
This tubing assembly is a stainless steel spare part, designed for high-pressure applications. It features specific internal and external diameters and is supplied with high pressure fittings, suitable for use in ACQUITY H-Class Systems.
  • Made from stainless steel.
  • Has an internal diameter of 0.004 in. (0.102 mm).
  • Measures 44.0 in. (1118 mm) in length.
  • Supplied with high pressure fittings.
  • Designed for UPLC systems.
  • Suitable for use in ACQUITY H-Class Systems.