Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Tubing Assembly, Stainless Steel, 0.004 in. ID, 6 in., High Pressure

Supplier:  Waters Corp 430002983

Encompass_Preferred

This replacement tubing assembly is designed for high-pressure applications. It is supplied with high pressure fittings and is compatible with ACQUITY H-Class Systems.

  • Supplied with high pressure fittings
  • Used in ACQUITY H-Class systems
  • Constructed from stainless steel
  • Internal diameter of 0.004 in.
  • Length of 6 in.
  • Suitable for UPLC systems

Catalog No. 50-239-2939


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.