Learn More
Waters Corp Tubing Assembly, Stainless Steel, 0.062 in. OD, 0.004 in. ID, 22.5 in. Length
Catalog No. 502390873 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This stainless steel tubing assembly is a high-pressure, critically clean component designed for use with UPLC systems, including H-Class Systems. It serves as a replacement part and is suitable for various laboratory applications requiring precise fluid transfer.
  • Made from stainless steel for durability.
  • Features an internal diameter of 0.004 in.
  • Has a length of 22.5 in.
  • Designed for UPLC system types.
  • Compatible with H-Class Systems.
  • Includes high-pressure fittings.
  • Provided as a single unit per package.
  • Serves as a spare part for various systems.
Catalog No. 50-239-0873 Supplier Waters Corp Supplier No. 430002717
This stainless steel tubing assembly is a high-pressure, critically clean component designed for use with UPLC systems, including H-Class Systems. It serves as a replacement part and is suitable for various laboratory applications requiring precise fluid transfer.
  • Made from stainless steel for durability.
  • Features an internal diameter of 0.004 in.
  • Has a length of 22.5 in.
  • Designed for UPLC system types.
  • Compatible with H-Class Systems.
  • Includes high-pressure fittings.
  • Provided as a single unit per package.
  • Serves as a spare part for various systems.