Learn More
Waters Corp Tubing Assembly, Stainless Steel, 0.007 in. ID, 19 in. Length
Catalog No. 502390465 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement tubing assembly is constructed from stainless steel. It is supplied with gold plated fittings and is designed for use on UPC2 systems.
  • Constructed from stainless steel.
  • Features an internal diameter of 0.007 in.
  • Measures 19 in. in length.
  • Equipped with gold plated fittings.
  • Designed for use with UPC2 systems.
  • Intended as a replacement part.
Catalog No. 50-239-0465 Supplier Waters Corp Supplier No. 430003215
This replacement tubing assembly is constructed from stainless steel. It is supplied with gold plated fittings and is designed for use on UPC2 systems.
  • Constructed from stainless steel.
  • Features an internal diameter of 0.007 in.
  • Measures 19 in. in length.
  • Equipped with gold plated fittings.
  • Designed for use with UPC2 systems.
  • Intended as a replacement part.