Learn More
Waters Corp Tubing Assembly, Stainless Steel, Pre-Heater to Column, Reusable
Catalog No. 502392744 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This reusable stainless steel tubing assembly is designed as a replacement part for connecting a pre-heater to a column, specifically associated with a CH-30A column heater.
  • Replacement tubing assembly
  • Made of stainless steel
  • Connects pre-heater to column
  • Reusable
  • Associated with a CH-30A column heater
  • Designed for UPLC systems
Catalog No. 50-239-2744 Supplier Waters Corp Supplier No. 700011766
This reusable stainless steel tubing assembly is designed as a replacement part for connecting a pre-heater to a column, specifically associated with a CH-30A column heater.
  • Replacement tubing assembly
  • Made of stainless steel
  • Connects pre-heater to column
  • Reusable
  • Associated with a CH-30A column heater
  • Designed for UPLC systems