Learn More
Waters Corp Tubing Assembly, Stainless Steel, Pump Head to Transducer, UPLC
Catalog No. 502390679 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed as a replacement part for connecting the pump head to the transducer. It is specifically used in ACQUITY Arc and Arc HPLC Quaternary Solvent Managers.
  • Constructed from stainless steel.
  • Serves as a tubing component for fluid transfer.
  • Used to connect the pump head to the transducer.
  • Compatible with ACQUITY Arc and Arc HPLC Quaternary Solvent Managers.
  • Intended for replacement as needed.
Catalog No. 50-239-0679 Supplier Waters Corp Supplier No. 700010679
This tubing assembly is designed as a replacement part for connecting the pump head to the transducer. It is specifically used in ACQUITY Arc and Arc HPLC Quaternary Solvent Managers.
  • Constructed from stainless steel.
  • Serves as a tubing component for fluid transfer.
  • Used to connect the pump head to the transducer.
  • Compatible with ACQUITY Arc and Arc HPLC Quaternary Solvent Managers.
  • Intended for replacement as needed.