Learn More
Waters Corp Tubing Assembly, Stainless Steel, UPLC, Transducer to Vent Valve
Catalog No. 502391195 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is a replacement part found on ACQUITY Arc and Arc HPLC Systems. It is constructed from stainless steel and connects the transducer to the vent valve.
  • Designed for UPLC systems.
  • Made from stainless steel.
  • Replacement part for transducer to vent valve connection.
  • Provided as 1 unit per package.
  • Replacement recommended as required.
Catalog No. 50-239-1195 Supplier Waters Corp Supplier No. 700010680
This tubing assembly is a replacement part found on ACQUITY Arc and Arc HPLC Systems. It is constructed from stainless steel and connects the transducer to the vent valve.
  • Designed for UPLC systems.
  • Made from stainless steel.
  • Replacement part for transducer to vent valve connection.
  • Provided as 1 unit per package.
  • Replacement recommended as required.