Learn More
Waters Corp Tubing Assembly, Stainless Steel, Transducer to Check Valve, UPC2
Catalog No. 502391523 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is a replacement part for a UPC2 Binary Solvent Manager. It is designed to connect the transducer to the check valve.
  • Made from stainless steel
  • Functions as a tubing assembly
  • Compatible with UPC2 system
  • Replace as required
Catalog No. 50-239-1523 Supplier Waters Corp Supplier No. 430003105
This tubing assembly is a replacement part for a UPC2 Binary Solvent Manager. It is designed to connect the transducer to the check valve.
  • Made from stainless steel
  • Functions as a tubing assembly
  • Compatible with UPC2 system
  • Replace as required