Learn More
Waters Corp Tubing Assembly, Stainless Steel, 0.007 in. ID, 9.50 in. Length
Catalog No. 502390896 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is a critical component for UPC2 Systems, crafted from durable stainless steel. It is designed to ensure reliable connections and performance within the system, featuring gold-plated fittings for enhanced conductivity and longevity.
  • Replacement tubing assembly
  • Supplied with gold plated fittings
  • Found on UPC2 systems
  • Made of stainless steel
  • Features a length of 9.50 in. (24.1 cm)
  • Has an internal diameter of 0.007 in.
  • Connects Port 5 of the sample manager injection valve to Port 4 of the convergence manager auxiliary valve
Catalog No. 50-239-0896 Supplier Waters Corp Supplier No. 430003350
This tubing assembly is a critical component for UPC2 Systems, crafted from durable stainless steel. It is designed to ensure reliable connections and performance within the system, featuring gold-plated fittings for enhanced conductivity and longevity.
  • Replacement tubing assembly
  • Supplied with gold plated fittings
  • Found on UPC2 systems
  • Made of stainless steel
  • Features a length of 9.50 in. (24.1 cm)
  • Has an internal diameter of 0.007 in.
  • Connects Port 5 of the sample manager injection valve to Port 4 of the convergence manager auxiliary valve