Learn More
Waters Corp Tubing Assembly, Stainless Steel, Vent Valve P2 to Mixer 2, UPLC
Catalog No. 502391186 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement tubing assembly is designed for use with ACQUITY Arc Systems.
  • Material: Stainless Steel
  • Configuration: Vent Valve P2 to Mixer 2
  • Application details: Found on ACQUITY Arc Systems
  • Product type: Spare Parts Kits
  • System type: UPLC
  • Units per package: 1 pk
  • Replacement schedule: As required
Catalog No. 50-239-1186 Supplier Waters Corp Supplier No. 700010686
This replacement tubing assembly is designed for use with ACQUITY Arc Systems.
  • Material: Stainless Steel
  • Configuration: Vent Valve P2 to Mixer 2
  • Application details: Found on ACQUITY Arc Systems
  • Product type: Spare Parts Kits
  • System type: UPLC
  • Units per package: 1 pk
  • Replacement schedule: As required