Learn More
Waters Corp Tubing Assembly, Transducer to Check Valve, MP35N
Catalog No. 502391037 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed for use in Bio ACQUITY Quaternary Solvent Managers and APC p-Isocratic Solvent Managers. It is constructed from MP35N for reliable performance.
  • Replacement tube assembly transducer to check valve tube.
  • Constructed from MP35N material.
  • Utilized on Bio ACQUITY Quaternary Solvent Managers.
  • Utilized on APC p-Isocratic Solvent Manager.
Catalog No. 50-239-1037 Supplier Waters Corp Supplier No. 430002475
This tubing assembly is designed for use in Bio ACQUITY Quaternary Solvent Managers and APC p-Isocratic Solvent Managers. It is constructed from MP35N for reliable performance.
  • Replacement tube assembly transducer to check valve tube.
  • Constructed from MP35N material.
  • Utilized on Bio ACQUITY Quaternary Solvent Managers.
  • Utilized on APC p-Isocratic Solvent Manager.