Learn More
Waters Corp Tubing Assembly, Union to Purge Valve
Catalog No. 502392896 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is specifically designed to connect a union to a purge valve. It is an essential spare part for maintaining system integrity and performance.
  • Designed to connect a union to a purge valve.
  • Identified as a spare part kit.
  • Compatible with ACQUITY detectors for ACQUITY arc system.
  • Suitable for the 2414 RI detector.
Catalog No. 50-239-2896 Supplier Waters Corp Supplier No. 700002364
This tubing assembly is specifically designed to connect a union to a purge valve. It is an essential spare part for maintaining system integrity and performance.
  • Designed to connect a union to a purge valve.
  • Identified as a spare part kit.
  • Compatible with ACQUITY detectors for ACQUITY arc system.
  • Suitable for the 2414 RI detector.