Learn More
Waters Corp Tubing Assembly, Vent Valve Port 1 to Waste, 0.010 in. ID
Catalog No. 502390812 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed for the ACQUITY BSM system, connecting the vent valve port 1 to waste with a 0.010 in. inner diameter.
  • Upgrade kit for ACQUITY BSM.
  • Includes one unit per package.
  • Replacement recommended as required.
Catalog No. 50-239-0812 Supplier Waters Corp Supplier No. 430001893
This tubing assembly is designed for the ACQUITY BSM system, connecting the vent valve port 1 to waste with a 0.010 in. inner diameter.
  • Upgrade kit for ACQUITY BSM.
  • Includes one unit per package.
  • Replacement recommended as required.