Learn More
Waters Corp Tubing Assembly, Vent Valve Port 4 to Waste, 1/16 in. OD x 0.010 in. ID
Catalog No. 502391969 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed for the ACQUITY BSM system. It connects the Vent Valve Port 4 to Waste, ensuring proper fluid management. The tubing features a 1/16 in. outer diameter and a 0.010 in. inner diameter.
  • Connects Vent Valve Port 4 to Waste.
  • Designed for ACQUITY BSM systems.
  • 1/16 in. outer diameter.
  • 0.010 in. inner diameter.
  • Supplied as 1 unit per package.
  • Replacement schedule is as required.
Catalog No. 50-239-1969 Supplier Waters Corp Supplier No. 430001894
This tubing assembly is designed for the ACQUITY BSM system. It connects the Vent Valve Port 4 to Waste, ensuring proper fluid management. The tubing features a 1/16 in. outer diameter and a 0.010 in. inner diameter.
  • Connects Vent Valve Port 4 to Waste.
  • Designed for ACQUITY BSM systems.
  • 1/16 in. outer diameter.
  • 0.010 in. inner diameter.
  • Supplied as 1 unit per package.
  • Replacement schedule is as required.