Learn More
Waters Corp Waters Tubing Assembly, GPV-A to Mixer Manifold, ACQUITY H-Class QSM
Catalog No. 50791974 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Tubing Assembly is a genuine replacement part specifically designed for the ACQUITY H-Class Quaternary Solvent Manager. It is used to address leaks from the tube leading from the GPV-A to the Mixer Manifold. Using genuine replacement parts ensures proper operation and longevity of equipment, as this assembly is made from the same material as the original tubing that came with the system. This part is crucial for upkeep but does not substitute for proper cleaning and maintenance.
  • Designed for ACQUITY H-Class Quaternary Solvent Manager
  • Addresses leaks in the GPV-A to Mixer Manifold tubing
  • Ensures proper equipment functioning
  • Made from original material
  • Replaces tubing in ACQUITY UPLC H-Class system
  • One unit per package
  • Replacement schedule as required
Catalog No. 50-791-974 Supplier Waters Corp Supplier No. 430002389
Add to Cart
Add to Cart
This Tubing Assembly is a genuine replacement part specifically designed for the ACQUITY H-Class Quaternary Solvent Manager. It is used to address leaks from the tube leading from the GPV-A to the Mixer Manifold. Using genuine replacement parts ensures proper operation and longevity of equipment, as this assembly is made from the same material as the original tubing that came with the system. This part is crucial for upkeep but does not substitute for proper cleaning and maintenance.
  • Designed for ACQUITY H-Class Quaternary Solvent Manager
  • Addresses leaks in the GPV-A to Mixer Manifold tubing
  • Ensures proper equipment functioning
  • Made from original material
  • Replaces tubing in ACQUITY UPLC H-Class system
  • One unit per package
  • Replacement schedule as required