Learn More
Waters Corp Waters Tubing Assembly, GPV-C to Mixer Manifold, ACQUITY UPLC H-Class QSM
Catalog No. 50791973 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed for the ACQUITY H-Class Quaternary Solvent Manager (QSM) and connects the GPV-C to the mixer manifold, ensuring reliable performance in UPLC systems.
  • Designed for ACQUITY UPLC Quaternary Solvent Manager.
  • Connects GPV-C to the mixer manifold.
  • Compatible with ACQUITY UPLC H-Class and H-Class PLUS Systems.
  • Suitable for UPLC system types.
  • Includes one unit per package.
  • Replacement is as required.
Catalog No. 50-791-973 Supplier Waters Corp Supplier No. 430002388
This tubing assembly is designed for the ACQUITY H-Class Quaternary Solvent Manager (QSM) and connects the GPV-C to the mixer manifold, ensuring reliable performance in UPLC systems.
  • Designed for ACQUITY UPLC Quaternary Solvent Manager.
  • Connects GPV-C to the mixer manifold.
  • Compatible with ACQUITY UPLC H-Class and H-Class PLUS Systems.
  • Suitable for UPLC system types.
  • Includes one unit per package.
  • Replacement is as required.