Learn More
Waters Corp WATERS CORPORATION Tubing Assembly, GPV-D to Mixer Manifold, UPLC, H-Class QSM
Catalog No. 50791972 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is engineered for use within the ACQUITY UPLC H-Class System, specifically facilitating the connection between the GPV-D and the mixer manifold in the ACQUITY H-Class QSM component.
  • Designed for ACQUITY UPLC QSM component.
  • Comes as 1 unit per package.
  • Tubing type for UPLC systems.
  • Compatible with ACQUITY UPLC H-Class System.
  • Recommended for replacement as required.
Catalog No. 50-791-972 Supplier Waters Corp Supplier No. 430002387
This tubing assembly is engineered for use within the ACQUITY UPLC H-Class System, specifically facilitating the connection between the GPV-D and the mixer manifold in the ACQUITY H-Class QSM component.
  • Designed for ACQUITY UPLC QSM component.
  • Comes as 1 unit per package.
  • Tubing type for UPLC systems.
  • Compatible with ACQUITY UPLC H-Class System.
  • Recommended for replacement as required.