Learn More
AVANTOR MASTERFLEX (VWR LLC) Tubing, Biopharm Plus, 2-stop, #14, 8/pk
Catalog No. NC1041504
Encompass
Masterflex Tubing, Biopharm Plus, 2-stop, #14, 8/pk, Ensure optimum performance from your Masterflex pumpUltra-smooth inner surface minimizes particle entrapment.Lower absorption; excellent biocompatibility; no leachable additive, DOP, or plasticizers.Very low extractables.Odorless and nontoxic, fungus-resistant.No taste imparted to transported fluids.
Catalog No. NC1041504 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9611614
Masterflex Tubing, Biopharm Plus, 2-stop, #14, 8/pk, Ensure optimum performance from your Masterflex pumpUltra-smooth inner surface minimizes particle entrapment.Lower absorption; excellent biocompatibility; no leachable additive, DOP, or plasticizers.Very low extractables.Odorless and nontoxic, fungus-resistant.No taste imparted to transported fluids.