Learn More
Formulatrix TUBING BLOCK (PACK OF 6)
SDP
Catalog No. NC2129623
Encompass
Tubing Block (Pack of 6).The tubing block is the fuidic connection between sample conical tubes and the Consumable Filtration Chip. This part works with any standard 50mL conical tube and features low-bind FEP tubing, (pack of 6).
This item is not returnable. View return policy
Catalog No. NC2129623 Supplier Formulatrix Supplier No. 795807A
Tubing Block (Pack of 6).The tubing block is the fuidic connection between sample conical tubes and the Consumable Filtration Chip. This part works with any standard 50mL conical tube and features low-bind FEP tubing, (pack of 6).