Learn More
New Age Industries Inc APSH Braid-Reinforced Silicone Hose, 3/8 in ID, 50 ft
SDP
Catalog No. NC1325001
Encompass_Preferred
Low volatile grade, platinum-cured silicone hose. Polyester yarn braiding inside the wall enhances the pressure capabilities.
Catalog No. NC1325001 Supplier New Age Industries Inc Supplier No. APSHP0375L
Low volatile grade, platinum-cured silicone hose. Polyester yarn braiding inside the wall enhances the pressure capabilities.