Learn More
AVANTOR MASTERFLEX (VWR LLC) C-Flex Tubing Braided; 3/4""id, 1 1/8""OD, 3/16"" Wall, 15'
Catalog No. NC1559236
Encompass
Applications: For use with cell cultures, vessel and tank transfer, pump applications and cold storage. Characteristics: C-Flex formula 082, clear, animal derived component free (ADCF) resin and reinforced with polyester braid. Certifications: Complies with USP Class VI standards. Temperature range: -50 to 275¿F (-45 to 135¿C) Sterilize: Autoclavable and gamma sterilizable.
Catalog No. NC1559236 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0643311
Applications: For use with cell cultures, vessel and tank transfer, pump applications and cold storage. Characteristics: C-Flex formula 082, clear, animal derived component free (ADCF) resin and reinforced with polyester braid. Certifications: Complies with USP Class VI standards. Temperature range: -50 to 275¿F (-45 to 135¿C) Sterilize: Autoclavable and gamma sterilizable.