Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Spooled High-Performance Precision Pump Tubing, C-Flex, L/S 24; 400 ft
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0642724/DEL
Encompass
Applications: General-purpose lab uses requiring high performance and long life Characteristics: Exceptional tensile and tear strength, good biocompatibility, low gas permeability, smooth surface, opaque white Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.
Catalog No. NC1039949