Learn More
Pacer Global LLC Saint-Gobain - 374 C-Flex TPE Tubing, 0.125'' ID, 0.25'' OD, 0.0625'' Wall, 50 Foot Coil, 1/EA - 374-125-2
SDP
Catalog No. 5000218261
Encompass_Preferred
Saint-Gobain - 374 C-Flex TPE Tubing, 0.125" ID, 0.25" OD, 0.0625" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-002-18261 Supplier Pacer Global LLC Supplier No. MLS3741252
Saint-Gobain - 374 C-Flex TPE Tubing, 0.125" ID, 0.25" OD, 0.0625" Wall, 50 Foot Coil, 1/EA