Learn More
Waters Corp Tubing, Capillary, BSM to MMV tee, 25µ x 24 in
Catalog No. 50791950 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The capillary tubing , 25µm x 24in long, fits between the BSM and the nano Tee in the 2D with Dilution configuration of the nanoACQUITY UPLC. To see the Fluidic Configurations for the nanoACQUITY UPLC, go to the Url Link below, under Systems select nanoACQUITY, click on the Waters Graphical Part Locator, and then choose Fluidic Configurations on the right side of the page. There will be the six different configurations of the nanoACQUITY that you can click on individually to see a larger image. There will also be a parts list for that specific configuration on the right side of the page.
Catalog No. 50-791-950 Supplier Waters Corp Supplier No. 430002140
The capillary tubing , 25µm x 24in long, fits between the BSM and the nano Tee in the 2D with Dilution configuration of the nanoACQUITY UPLC. To see the Fluidic Configurations for the nanoACQUITY UPLC, go to the Url Link below, under Systems select nanoACQUITY, click on the Waters Graphical Part Locator, and then choose Fluidic Configurations on the right side of the page. There will be the six different configurations of the nanoACQUITY that you can click on individually to see a larger image. There will also be a parts list for that specific configuration on the right side of the page.