Learn More
Waters Corp Tubing Assembly, Capillary with frit, 25µm x 22 in
Catalog No. 50791953 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Capillary Tubing connects from the second BSM to the inject valve on the nanoACQUITY UPLC in the following configurations: Vented Dual Pump Configuration, Dual Pump Trapping Configuration, 2D with Dilution Configuration, and 2D Configuration. To see the Fluidic Configurations for the nanoACQUITY UPLC, go to the Url Link below, under Systems select nanoACQUITY, click on the Waters Graphical Part Locator, and then choose Fluidic Configurations on the right side of the page. There will be the six different configurations of the nanoACQUITY that you can click on individually to see a larger image. There will also be a parts list for that specific configuration on the right side of the page.
Catalog No. 50-791-953 Supplier Waters Corp Supplier No. 430002155
This Capillary Tubing connects from the second BSM to the inject valve on the nanoACQUITY UPLC in the following configurations: Vented Dual Pump Configuration, Dual Pump Trapping Configuration, 2D with Dilution Configuration, and 2D Configuration. To see the Fluidic Configurations for the nanoACQUITY UPLC, go to the Url Link below, under Systems select nanoACQUITY, click on the Waters Graphical Part Locator, and then choose Fluidic Configurations on the right side of the page. There will be the six different configurations of the nanoACQUITY that you can click on individually to see a larger image. There will also be a parts list for that specific configuration on the right side of the page.