Learn More
Microsolv Technology Corporation Tubing, Capillary, PEEK, 75um ID, 360um OD, Natural Color. 10 Meter Roll. 1 EA.
Catalog No. 502271475
Encompass_Preferred
This small OD, PEEK, capillary tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives. The internal and outer dimensional tolerances of this tubing is +/- 0.05mm. Color: Natural. Pressure rating: 5200 psi.
This item is not returnable. View return policy
Catalog No. 50-227-1475 Supplier Microsolv Technology Corporation Supplier No. 4731005
This small OD, PEEK, capillary tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives. The internal and outer dimensional tolerances of this tubing is +/- 0.05mm. Color: Natural. Pressure rating: 5200 psi.