Learn More
Waters Corp Waters Tubing Assembly, Capillary, With Frit, 25 μm, 12 in.
Catalog No. 50791952 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This capillary tubing connects from the trap valve to the MMV tee on the nanoACQUITY UPLC in the 2D with Dilution Configuration.
  • Capillary tubing design
  • Connects from trap valve to MMV tee on the nanoACQUITY UPLC
  • Suitable for 2D with dilution configuration
  • Dimensions: 25μm x 12 in.
  • Supplied as 1 pack
  • Replacement schedule: As required
Catalog No. 50-791-952 Supplier Waters Corp Supplier No. 430002153
This capillary tubing connects from the trap valve to the MMV tee on the nanoACQUITY UPLC in the 2D with Dilution Configuration.
  • Capillary tubing design
  • Connects from trap valve to MMV tee on the nanoACQUITY UPLC
  • Suitable for 2D with dilution configuration
  • Dimensions: 25μm x 12 in.
  • Supplied as 1 pack
  • Replacement schedule: As required