Learn More
Waters Corp Waters Tubing Assembly, Capillary, 75μm, 10 in., with Frit
Catalog No. 50791940 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This capillary tubing assembly, featuring a frit, is designed for various laboratory applications. It measures 75μm in diameter and 10 in. in length, providing precise fluid transfer. This assembly is also an integral component of the Mixer Kit for 1mm Columns (part number 205000540).
  • Capillary tubing with frit
  • Measures 75μm in diameter
  • 10 in. length
  • Supplied as 1 unit per package
  • Suitable for replacement as required
Catalog No. 50-791-940 Supplier Waters Corp Supplier No. 430001837
This capillary tubing assembly, featuring a frit, is designed for various laboratory applications. It measures 75μm in diameter and 10 in. in length, providing precise fluid transfer. This assembly is also an integral component of the Mixer Kit for 1mm Columns (part number 205000540).
  • Capillary tubing with frit
  • Measures 75μm in diameter
  • 10 in. length
  • Supplied as 1 unit per package
  • Suitable for replacement as required