Learn More
Chemglass Life Sciences Tubing Clamp, 1/2" OD, Single Position
SDP
Catalog No. 5000083242
Encompass_Preferred
Plastic clamp is manufactured from White Polypropylene and is both autoclavable and gamma stable.Clamp is designed for on/off control of fluid or air.One-hand control and economically priced for disposable applications.Packed 25 per Case.
Catalog No. 50-000-83242 Supplier Chemglass Life Sciences Supplier No. CLS1436SP5
Plastic clamp is manufactured from White Polypropylene and is both autoclavable and gamma stable.Clamp is designed for on/off control of fluid or air.One-hand control and economically priced for disposable applications.Packed 25 per Case.