Learn More
Research Products International Corp Tubing Clamp, 5/16-7/8"", 10/pk
SDP
Catalog No. 501978416
Encompass
So easy to install, it's no wonder these are the most frequently used style of hose clamps. Clamps are for firm plastic and rubber hose and tube. Do not exceed the maximum torque or clamps may be damaged. Stainless steel offers good corrosion resistance. When choosing a clamp, measure the outside diameter of your hose or tube with the fitting installed.
Catalog No. 50-197-8416 Supplier Research Products International Corp Supplier No. 255505
So easy to install, it's no wonder these are the most frequently used style of hose clamps. Clamps are for firm plastic and rubber hose and tube. Do not exceed the maximum torque or clamps may be damaged. Stainless steel offers good corrosion resistance. When choosing a clamp, measure the outside diameter of your hose or tube with the fitting installed.