Learn More
Cole-Parmer Keck Ramp Clamp Tubing Clamps, 10 mm, 12/Pk
Supplier:  Cole-Parmer 0683507
Encompass_Preferred
Unique wheel-in-groove clamp design permits one-handed flow control. Simply remove wheel to insert tubing, then roll wheel back into clamp to your desired flow rate. Made of pure PBT, these clamps are safe to 392°F (200°C). All four sizes accept tubing with a maximum wall thickness of 3/32" (2.4 mm).
Catalog No. NC9647758