Learn More
Hach Company Tubing, Tygon, E-3603, 3.2 mm (I.D.)
Supplier:  Hach Company 2074038
Encompass
Chemical-resistant, Formula E-3603 Tygon tubing with flex strength greater than rubber. Unaffected by oxidizing agents and compatible with virtually any laboratory chemical. Bends readily to sharp radius curves and will maintain strong grip on glass or metal. Highly polished bore prevents interior build-up while facilitating cleaning.
Catalog No. 50-199-5222