Learn More
Waters Corp WATERS CORPORATION Ferrule, Flangeless, Tefzel, 3.0 mm ID
Catalog No. 50792910 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a flangeless ferrule, precisely engineered from Tefzel, designed to establish secure connections for 3.0 mm inner diameter tubing. Functioning as an inlet end nut, it is an essential component for maintaining system integrity in various laboratory applications, particularly within Waters ACQUITY UPLC and ACQUITY H-Class PLUS Bio systems.
  • Flangeless design for secure connections.
  • Constructed from Tefzel material.
  • Designed for 3.0 mm ID tubing.
  • Includes a lock ring for enhanced sealing.
  • Functions as an inlet end nut.
  • Compatible with ACQUITY UPLC and ACQUITY H-Class PLUS Bio systems.
Catalog No. 50-792-910 Supplier Waters Corp Supplier No. 700003792
This product is a flangeless ferrule, precisely engineered from Tefzel, designed to establish secure connections for 3.0 mm inner diameter tubing. Functioning as an inlet end nut, it is an essential component for maintaining system integrity in various laboratory applications, particularly within Waters ACQUITY UPLC and ACQUITY H-Class PLUS Bio systems.
  • Flangeless design for secure connections.
  • Constructed from Tefzel material.
  • Designed for 3.0 mm ID tubing.
  • Includes a lock ring for enhanced sealing.
  • Functions as an inlet end nut.
  • Compatible with ACQUITY UPLC and ACQUITY H-Class PLUS Bio systems.