Learn More
Med Vet International McKesson LUMEON™ Blood Pressure Unit Tubing, 01-885NGM, 1/Pack
Catalog No. 503045542
Encompass_Preferred
Blood Pressure Tubing McKesson LUMEON™ Includes Male Luer Connector (01-891MGM) For use with most Aneroid Sphygmomanometers Facilitates blood pressure measurement when using wall or mobile instruments Coiled Tubing, Black, 8-Foot When Extended 2-year warranty
Catalog No. 50-304-5542 Supplier Med Vet International Supplier No. 1029662PK
Blood Pressure Tubing McKesson LUMEON™ Includes Male Luer Connector (01-891MGM) For use with most Aneroid Sphygmomanometers Facilitates blood pressure measurement when using wall or mobile instruments Coiled Tubing, Black, 8-Foot When Extended 2-year warranty