Learn More
Med Vet International Medi-Vac Suction Tubing, 3/16 Inch Inner Diameter, 6 Foot Length, N56A, 50/Case
Catalog No. 503044915
Encompass_Preferred
Resists collapse from pressure, up to 22 Hg vacuum Single use, sterile Maxi-Grip™ connectors Rigid male connector
Catalog No. 50-304-4915 Supplier Med Vet International Supplier No. 251161CS
Resists collapse from pressure, up to 22 Hg vacuum Single use, sterile Maxi-Grip™ connectors Rigid male connector